D-(+)-Tryptophan ≥99%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210314425 0210314480
IC10314425EA 244.41 USD
IC10314425 IC10314480
D-(+)-Tryptophan ≥99%, Powder
D-(+)-Tryptophan

D-Tryptophan is an amino acid and a unnatural isomer of tryptophan.


Soluble in water or hot alcohol. Also soluble in 0.5M Hydrochloric Acid (50 mg plus 1 mL 0.5M HCl) giving clear to slightly hazy solution.

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD00005647
CAS Number: 153-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR