Triethanolamine hydrochloride ≥99%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210310980 0210310983
IC10310980EA 51.25 USD
IC10310980 IC10310983
Triethanolamine hydrochloride ≥99%, Powder
Triethanolamine hydrochloride

Triethanolamine is a zwitterionic buffer.


Triethanolamine(TEA) is used primarily as an emulsifier and surfactant. It neutralizes fatty acids, adjusts and buffers the pH, and solubilises oils and other ingredients that are not completely soluble in water. Triethanolamine is also used as organic additive (0.1 wt. %) in the grinding of cement clinker. Another common use of TEA is as a complexing agent for aluminium ions in aqueous solutions.


Soluble in water (30% w/v - clear, colorless solution); miscible in methanol, acetone, benzene, ether.


Triethanolamine is a zwitterionic buffer.

Formula: C₆H₁₆ClNO₃
MW: 185.65 g/mol
MDL Number: MFCD00012596
CAS Number: 637-39-8

Order Now

Specification Test Results

Synonyms 2,2,2-Nitrilotriethanol hydrochloride; Tris(2-hydroxyethyl)amine hydrochloride
Form White Powder
Storage Store at Room Temperature(15-30 °C), desiccate.
pH 4.0 - 5.5(1% aq soln), useful pH range 7.3 - 8.3
pka 7.8(25 °C) (Lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR