DL-Threonine ≥99%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210305010 0210305025 0210305080 0210305091
IC10305010EA 39.26 USD
IC10305010 IC10305025 IC10305080 IC10305091
DL-Threonine ≥99%, white powder
DL-Threonine

DL-Threonine is an essential α - amino acid classified as polar which has two chiral centers. In plants and micro-organisms, threonine is synthesized from aspartic acid via aspartyl - semialdehyde and homoserine.


DL-Threonine is as an additive to animal feed. They are also used to chelate metal cations in order to improve the absorption of minerals from supplements, which may be required to improve the health or production of animals. This chelating ability is also used in fertilizers for agriculture.


Soluble in water (50 mg/mL- clear, colorless solution).


DL-Threonine is an essential α - amino acid classified as polar which has two chiral centers. In plants and micro-organisms, threonine is sunthesized from aspartic acid via aspartyl - semialdehyde and homoserine.

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 238…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00063722
CAS Number: 80-68-2

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Loss on Drying ≤0.2%
Optical Rotation -1° to +1° (c=3, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR