Thiamine hydrochloride, powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.
Thiamine hydrochloride is used as a food-additive to add a brothy/meaty flavor to gravies or soups. It is also used as food supplement
Soluble in HCl (50 mg/mL-clear, colorless solution).
Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.
Precaución: Research Use Only (RUO).
Formula:
C₁₂H₁₈Cl₂N₄OS MW: 337.27 g/mol Melting Pt: 252 Storage Temperature: Ambient |
MDL Number:
MFCD00012780 CAS Number: 67-03-8 Merck Index: 13,09366 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
pH (1% aq soln) | 2,7 - 3,4 |
OD400 (1% aq soln) | ≤0.25 |
Loss on Drying | ≤5,0% |
Residue on Ignition | ≤0,2% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...