Theophyllin, anhydrous, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline

0210302480 0210302483 0210302490 0210302491
IC10302480EA 61.28 USD
IC10302480 IC10302483 IC10302490 IC10302491
Theophyllin, anhydrous, white powder
Theophyllin

Phosphodiesterase inhibitor; diuretic; cardiac stimulant; muscle relaxant; asthma medication.


Soluble in 0.1 M HCl, 0.1 M NaOH; Slightly soluble in water; moderately soluble in ethanol. Soluble in 1 M NH4OH (50 mg/mL - clear colorless solution) and 50% ammonium hydorxide (50 mg/mL - clear, colorless solution).


Phosphodiesterase inhibitor; diuretic; cardiac stimulant; muscle relaxant; asthma medication.


Precaución: Research Use Only (RUO).

Formula: C₇H₈N₄O₂
MW: 180.16 g/mol
MDL Number: MFCD00079619
CAS Number: 58-55-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR