Tetracycline hydrochloride, yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0210301105 0210301125 0210301180
IC10301105EA 59.63 USD
IC10301105 IC10301125 IC10301180
Tetracycline hydrochloride, yellow powder
Tetracycline hydrochloride

Tetracycline is a broad spectrum polyketide antibiotic with clinical uses in treating bacterial infections such as Rocky Mountain spotted fever, typush fever, tick fevers, Q fever, and Brill-Zinsser disease and to treat upper respiratory infections and acne. It has been used in studies of multidrug resistance and potential side effects including acute pancreatitis.


Tetracyclines possess a wide range of antimicrobial activity against gram-positive and gram-negative bacteria. The bacterial ribosome is the site of action of tetracyclines. Access to the ribosomes of gramnegative bacteria is obtained by passive diffusion through hydrophilic pores in the outer cell membrane and then by an energy-dependent active transport system that pumps all tetracyclines through the inner cytoplasmic membrane. This active transport system may require a periplasmic protein carrier. Tetracyclines bind specifically to 30S ribosomes and appear to inhibit protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. The inhibitory effects of the tetracyclines can be reversed by washing. This suggests that the reversibly bound antibiotic rather than the small portion of irreversibly bound drug is responsible for the antibacterial action.


Tetracyline Hydrochloride acts as an inhibitor of protein synthesis by binding to the 30S subunit and inhibiting binding of aminoacyl-tRNA. Antibiotic substance produced by Streptomyces spp.


Tetracycline hydrochloride is used in tetracycline controlled gene expression systems (gene switches) such a the tet-on and tet-off systems.


Freely soluble in water or 0.1 N HCl (10 mg/ml - clear, bright yellow solution); soluble in methanol, ethanol. Insoluble in ether, hydrocarbons. pH (2% aq soln): ~2.1-2.3.


Tetracyclines possess a wide range of antimicrobial activity against gram-positive and gram-negative bacteria. The bacterial ribosome is the site of action of tetracyclines. Access to the ribosomes of gramnegative bacteria is obtained by passive diffusion through hydrophilic pores in the outer cell membrane and then by an energy-dependent active transport system that pumps all tetracyclines through the inner cytoplasmic membrane. This active transport system may require a periplasmic protein carrier. Tetracyclines bind specifically to 30S ribosomes and appear to inhibit protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. The inhibitory effects of the tetracyclines can be reversed by washing. This suggests that the reversibly bound antibiotic rather than the small portion of irreversibly bound drug is responsible for the antibacterial action.


Precaución: Research Use Only (RUO).

Formula: C₂₂H₂₅ClN₂O₈
MW: 480.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078142
CAS Number: 64-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=340c391983b091bb27a8d0e5b217246c40d5a8ec&vid=340c391983b091bb27a8d0e5b217246c40d5a8ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Potency ≥900 µg/mg
FTIR Conforms to Standard
Melting Point 215 - 225 °C
UV/Vis: λmax (1N HCl) 219, 271, 357 ± 5 nm
pH (1% aq soln) 1.8 - 2.8
Moisture Content ≤2%
Optical Rotation -239 ° to -258 ° (c=0.01, 0.1 N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR