Starch, soluble (from rice), white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Starch, Vitex® iodine indicator, Amylodextrin
Starch can be hydrolyzed into simpler carbohydrates by acids, various enzymes, or a combination of the two and can be used as a bland-tasting filler, thickener, sweetner and in hydrogen production.
Starch becomes soluble in water when heated.
Starch or amylum is a carbohydrate consisting of a large number of glucose units joined by glycosidic bonds. Starch becomes soluble in water when heated. The granules swell and burst, the semi-crystalline structure is lost and the smaller amylose molecules start leaching out of the granule, forming a network that holds water and increasing the mixture's viscosity. This process is called starch gelatinization. During cooking the starch becomes a paste and increases further in viscosity. During cooling or prolonged storage of the paste, the semi-crystalline structure partially recovers and the starch paste thickens, expelling water. This is mainly caused by retrogradation of the amylose.
Precaución: Research Use Only (RUO).
Formula:
(C₆H₁₀O₅)n MW: 162.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS Number: 9005-84-9 Merck Index: 13,08878 |
Specification Test Results
Identity Test | Passes |
Source | Rice |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...