Sorbic acid ≥99%, yellow crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
(E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
Sorbic acid (2,4-hexadienoic acid) is a straight chain unsaturated fatty acid. It is an antimicrobial agent. The antimicrobial action of sorbic acid is due to its inhibitory influence on various enzymes in the microbial cell such as enzymes involved in carbohydrate metabolism such as enolase and lactate dehydrogenase, enzymes of citric acid cycles.
Sorbic acid is an antimicrobial agents often used as preservatives in food and drinks to prevent the growth of mold, yeast, and fungi.
Sorbic acid can be used to inhibit bacterial, yeast and fungal sulfhydryl enzymes by inhibiting amino acid uptake.
Moderately soluble in water.
Sorbic acid (2,4-hexadienoic acid) is a straight chain unsaturated fatty acid. It is an antimicrobial agent. The antimicrobial action of sorbic acid is due to its inhibitory influence on various enzymes in the microbial cell such as enzymes involved in carbohydrate metabolism such as enolase and lactate dehydrogenase, enzymes of citric acid cycles.
Precaución: Research Use Only (RUO).
Formula:
CH₃CH=CHCH=CHCOOH MW: 112.13 g/mol Boiling Pt: >190 °C (1013 hPa) (lit.) Melting Pt: 130…140 °C Density: 1.2 g/cm³ (20 °C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00002703 CAS Number: 110-44-1 |
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
Melting Point | 130 - 140 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...