Starch, soluble (from potato), powder, Reagent Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Starch, Vitex® iodine indicator, Amylodextrin
0210293590
0210293591
IC10293590EA
114.54
USD
IC10293590
IC10293591
Starch, soluble (from potato), powder, Reagent Grade
Starch, soluble
2% solution in boiling water.
Soluble as 2% solution in boiling water.
Precaución: Research Use Only (RUO).
Formula:
(C₆H₁₀O₅)n MW: 162.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS Number: 9005-84-9 |
Specification Test Results
Identity Test | Passes |
Source | Potato |
FTIR | Conforms to Standard |
Moisture Content | <20% |
pH (2% aq soln) | 5 - 7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...