Sodium mercaptoacetate (sodium thioglycolate) ≥97% for bacteriology
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Thioglycolic Acid, Thioglycolic Acid Sodium Salt, Mercaptoacetic acid, sodium salt
Thioglycolic acid protects tryptophan in amino acid analysis,and also mediates formation of ATP from ADP. It lowers the oxidation-reduction potential and neutralises mercurial preservatives. An inhibitor of fatty acid oxidation. An agent that prevents the metabolism of fatty acids and stimulates feeding. The sodium salt form is typically used in production of bacteriological culture media. The free acid is used as a reagent for the sensitive detection of iron (a blue color appears in the presence of ferric iron, and when an alkali hydroxide is added to a solution containing ferrous salts and thioglycolic acid, a yellow precipitate forms), molybdenum, silver and tin; used for the extraction and spectrophotometric determination of various transition metals such as lead, tungsten, molybdenum and titanium.
Sodium thioglycolate is a commonly used reagent for bacteriological research to maintain reducing conditions in media. Thioglycolate can also protect enzymes against inactivation by maintaining protein thiol groups in the reduced state. Thioglycolate medium is frequently used in inflammation research to elicit a neutrophil and macrophage response in vivo.
(200 mg/1 mL water): Clear Solution
Thioglycolic acid protects tryptophan in amino acid analysis,and also mediates formation of ATP from ADP. It lowers the oxidation-reduction potential and neutralises mercurial preservatives. An inhibitor of fatty acid oxidation. An agent that prevents the metabolism of fatty acids and stimulates feeding. The sodium salt form is typically used in production of bacteriological culture media. The free acid is used as a reagent for the sensitive detection of iron (a blue color appears in the presence of ferric iron, and when an alkali hydroxide is added to a solution containing ferrous salts and thioglycolic acid, a yellow precipitate forms), molybdenum, silver and tin; used for the extraction and spectrophotometric determination of various transition metals such as lead, tungsten, molybdenum and titanium.
Formula:
HSCH₂COONa MW: 114.1 g/mol |
MDL Number:
MFCD00043386 CAS Number: 367-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...