Taurocholic acid sodium salt ≥97%, powder, Practical grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Cholaic acid sodium salt, Taurocholic acid sodium, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate (1:1), Sodium taurocholate
Applications
Taurocholic Acid Sodium Salt is an anionic detergent used for protein solubilization.
Product Description
Taurocholic Acid Sodium Salt is the conjugation product of cholic acid with taurine and the principal constituent of the bile of carnivorous animals.
Biochem/physiol Actions
Non-sulfated bile salt, physiological transport substrate for the bile salt export pump/sister of Pgp (BSEP/spgp), Na(+)/taurocholate cotransporter (NTCP) and MRP3 into the hepatic carnalicular.
Melting Point: 230 °C (decomposes)(Lit.)
Presentation: White Powder
Format: Powder
pH: 6 - 7.5(1% aq soln)
Aggregation Number: 4
Critical Micelle Concentration (CMC): 3-11 mM
Heavy Metals: =20 ppm
Loss on Drying: =7.0%
NOTES: Sodium Cholate : =3.0%
Solubility: Soluble in water (100 mg/mL).
Storage & Handling: Store at Room Temperature (15-30 °C).
Formula:
C₂₆H₄₄NNaO₇S MW: 537.688 g/mol |
MDL Number:
MFCD00013050 CAS Number: 145-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...