di-Sodium succinate hexahydrate ≥98%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0210292980 0210292990 0210292991
IC10292980EA 38.63 USD
IC10292980 IC10292990 IC10292991
di-Sodium succinate hexahydrate ≥98%, white crystalline powder

Soluble in water (100 mg/mL - clear, colorless solution).


Use as a TCA (Krebs cycle) supplement in cell culture applications. Sodium succinate dibasic hexahydrate can be used to study x-ray crystallography, protein structural analysis, proteomics and optimization reagents. Sodium succinate dibasic hexahydrate has been used to determine the calorimetric and diffractometric evidence for the sequential crystallization of buffer components and the consequential pH swing in frozen solutions. Sodium succinate dibasic hexahydrate has also been used to provide insights into the mechanism of Pseudomonas dacunhae aspartate ß-decarboxylase from rapid-scanning stopped-flow kinetics.


Soluble in water (100 mg/mL - clear, colorless solution).

Formula: NaOOCCH₂CH₂COONa·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149117
CAS Number: 6106-21-4

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (1% aq soln) 7.7 - 8.7
Moisture Content 35 - 40%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR