Sodium diatrizoate ≥99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Diatrizoate sodium, 3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid sodium salt, 3,5-Diacetamido-2,4,6-triiodobenzoic acid sodium salt, Diatrizoic acid sodium salt, Amidotrizoate sodium salt, Sodium 3,5-diacetamido-2,4,6-triiodobenzoate

0210290810 0210290850 0210290880
IC10290810EA 37.28 USD
IC10290810 IC10290850 IC10290880
Sodium diatrizoate ≥99%, white crystalline powder
Sodium diatrizoate

Sodium Diatrizoate has been shown to cause partial redistribution of tight-junction-associated membrane proteins into a cytoplasmic compartment in cultured monolayer MDCK cells.


Sodium diatrizoate is used as a density gradient reagent in blood cell separation. It has also been utilized as a radiological contrast agent.


Soluble in water (350 mg/mL).


Sodium Diatrizoate has been shown to cause partial redistribution of tight-junction-associated membrane proteins into a cytoplasmic compartment in cultured monolayer MDCK cells.


Precaución: Research Use Only (RUO).

Formula: C₁₁H₈I₃N₂NaO₄
MW: 635.898 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051544
CAS Number: 737-31-5

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
pH (10% solution) 7.0 - 9.7

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR