Sodium L(+)-ascorbate ≥99%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt

0210289025 0210289090 0210289091 0210289080
IC10289025EA 74.35 USD
IC10289025 IC10289090 IC10289091 76102-772
Sodium L(+)-ascorbate ≥99%, white powder
Sodium L(+)-ascorbate

L-Ascorbic Acid, Sodium Salt is a vitamin widely distributed in the plant and animal kingdom.


  • pKa: pK1: 4.17, pK2: 11.57(Lit.)
  • Oxidizes on exposure to light and air


L-Ascorbic Acid, Sodium Salt is used as an antimicrobial and antioxidant in foodstuffs, a reductant for the isolation of chloroplasts. It has an effect on the nonspecific immune system. It preserves nitric oxide (NO) which may help prevent endothelial dysfunction, helps prevent and reverse the oxidative damage done to proteins and lipids as a result of cigarette smoke.


L-Ascorbic acid (Vitamin C) is a water soluble molecule use in a wide variety of applications, including cell culture, as a reducing agent that helps reduce oxidative stress. L-Ascorbate can be regenerated by biological systems.


Soluble in water (62 g/100 ml at 25 °C; 78 g/100 mL at 75 °C)


L-Ascorbic Acid, Sodium Salt is used as an antimicrobial and antioxidant in foodstuffs, a reductant for the isolation of chloroplasts. It has an effect on the nonspecific immune system. It preserves nitric oxide (NO) which may help prevent endothelial dysfunction, helps prevent and reverse the oxidative damage done to proteins and lipids as a result of cigarette smoke.

Formula: C₆H₇NaO₆
Melting Pt: 213…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00082340
CAS Number: 134-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d142b281353c28f2a72502b60fece03e926023d&vid=988032859afe97984b0fc1b8bfd43cff093f099a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Optical Rotation (c=1, water) +100° to +110°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR