Safflower oil, Food Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Carthamus tinctorius oil, Triglyceride of Fatty Acids, Fats and Glyceridic oils, safflower, Oils, glyceridic, safflower, Thistle oil, Safflower seed oil

0210288890 0210288891
IC10288890EA 105.84 USD
IC10288890 IC10288891
Safflower oil, Food Grade
Safflower oil

Linoleic Acid 70-80%


Safflower oil is used for preventing heart disease, including "hardening of the arteries" (atherosclerosis) and stroke. It is also used to treat fever, tumors, coughs, breathing problems, clotting conditions, pain, heart disease, chest pain, and traumatic injuries. In foods, safflower seed oil is used as a cooking oil. In manufacturing, safflower flower is used to color cosmetics and dye fabrics. Safflower seed oil is used as a paint solvent.

MDL Number: MFCD00132216
CAS Number: 8001-23-8

Order Now

Specification Test Results

Synonyms Carthamustinctorius oil; Diestel oil; Diestelol; Distelol; Safflower seed oil ;Saffola; Thistle oil
Concentration 70-80% Linoleic Acid
Form Oil
Storage Store at Room Temperature (15-30 °C).
Refractive Index n20/D 1.476 (lit.)
Source Carthamus tinctorius seed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR