β-Sitosterol, white powder, Practical grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: α-Dihydrofucosterol, β-Sitosterol, 5-Stigmasten-3β-ol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol

0210288610 0210288680 0210288690
IC10288610EA 56.77 USD
IC10288610 IC10288680 IC10288690
β-Sitosterol, white powder, Practical grade
beta-Sitosterol

β-Sitosterol has recently been shown to induce G2/M arrest, endoreduplication, and apoptosis through the Bcl-2 and PI3K/Akt signaling pathways.


Soluble in chloroform (50 mg/ml - clear, colorless solution).


β-Sitosterol has recently been shown to induce G2/M arrest, endoreduplication, and apoptosis through the Bcl-2 and PI3K/Akt signaling pathways.


Precaución: Research Use Only (RUO).

Formula: C₂₉H₅₀O
MW: 414.72 g/mol
MDL Number: MFCD00003631
CAS Number: 83-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cf6ec423fe16d3ddf68843864a7ca11bcfcacdb&vid=7cf6ec423fe16d3ddf68843864a7ca11bcfcacdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Source Soybeans
beta-Sitosterol 40 - 60%
Campesterol 20 - 40%
Stigmasterol ≥5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR