DL-Serine ≥98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid

0210286825 0210286880 0210286891
IC10286825EA 63.95 USD
IC10286825 IC10286880 IC10286891
DL-Serine ≥98%, white powder
DL-Serine

NMDA agonist acting at the glycine site; precursor of glycine by serine hydroxymethyltransferase.


Soluble in water (50 mg/mL-clear colorless solution).


Precaución: Research Use Only (RUO).

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 395
Melting Pt: 228 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064223
CAS Number: 302-84-1
Merck Index: 13,08534

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Melting Point 234 - 244 °C
Optical Rotation (c = 5, 1N HCl) -1° to + 1°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR