Rubidium chloride ≥99%, white powder, Optical Grade Optical Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Rubidiumchloride

0210282505 0210282510 0210282550
IC10282505EA 169.97 USD
IC10282505 IC10282510 IC10282550
Rubidium chloride ≥99%, white powder, Optical Grade Optical Grade
Rubidium chloride

Rubidium chloride is used as a catalyst and additive in gasoline.


In materials science, RbCl has been used to prepare molecular nanowires as potential precursors of nanoscale devices. In chromatography, RbCl and other univalent salts have been studied for their influence on the capillary electrophoretic separation of amino acids labeled with 3-(4-carboxybenzoyl)-quinoline-2-carboxaldehyde. RbCl is utilized in the transfection of bacteria. RbCl has been used to investigate the gating and permeability of ion channels produced by botulinum neurotoxin types A and E in membranes from cultured PC12 cells.


Rubidium chloride is used as a catalyst and additive in gasoline. In materials science, RbCl has been used to prepare molecular nanowires as potential precursors of nanoscale devices. In chromatography, RbCl and other univalent salts have been studied for their influence on the capillary electrophoretic separation of amino acids labeled with 3-(4-carboxybenzoyl)-quinoline-2-carboxaldehyde. RbCl is utilized in the transfection of bacteria. RbCl has been used to investigate the gating and permeability of ion channels produced by botulinum neurotoxin types A and E in membranes from cultured PC12 cells.

Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390
Melting Pt: 715 (lit.)
MDL Number: MFCD00011187
CAS Number: 7791-11-9

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
OD280 (40% aq soln) ≤0.2
Solubility (40% aq soln) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR