D-(+)-Raffinose pentahydrate ≥98%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate

0210279725 0210279780 0210279790 0210279750
IC10279725EA 91.08 USD
IC10279725 IC10279780 IC10279790 75875-596
D-(+)-Raffinose pentahydrate ≥98%, white crystalline powder
D-(+)-Raffinose pentahydrate

Raffinose is a trisaccharide composed of galactose, fructose, and glucose.


Raffinose is hydrolysed to D-galactose and sucrose by D-galactosidase.


Soluble in pyridine and water (1 g/7 ml)

Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
Melting Pt: 76…86 °C
Density: 1.465 g/mL (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00071590
CAS Number: 17629-30-0
EINECS: 208-146-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4145244c500acd6de21c3fe6398c844f1b27503&vid=d4145244c500acd6de21c3fe6398c844f1b27503&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Melting Point 76 - 86°C
Optical Rotation (c=4, water) 101 - 107°
Loss on Drying 13 - 17%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR