3,5-Dihydroxytoluene monohydrate ≥98%, light beige powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 5-Methylresorcinol monohydrate, Orcinol monohydrate, 5-Methylbenzene-1,3-diol hydrate

0210250610 0210250650 0210250605
IC10250610EA 73.1 USD
IC10250610 IC10250650 76204-362
3,5-Dihydroxytoluene monohydrate ≥98%, light beige powder
3,5-Dihydroxytoluene monohydrate

Orcinol is used to prepare colorimetric reagents for the determination of RNA, DNA, hexoses and pentoses.


Solubility : Soluble in alcohol and water (5 mg/ mL-clear colorless to faint yellow solution).


Orcinol is used to prepare colorimetric reagents for the determination of RNA, DNA, hexoses and pentoses.


Precaución: Storage: protect from light. Research Use Only (RUO).

Formula: C₇H₈O₂·1H₂O
MW: 142.15 g/mol
Melting Pt: 57...67 °C
Storage Temperature: Ambient
MDL Number: MFCD00149092
CAS Number: 6153-39-5

Order Now

Specification Test Results

Identity Test Passes
Purity 98%
FTIR Conforms to Standard
Melting Point 57 - 67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR