4-Nitrophenol ≥99% (by GC), crystalline powder for spectroscopy
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0210246101
0210246105
IC10246101EA
56.63
USD
IC10246101
IC0210246105
4-Nitrophenol ≥99% (by GC), crystalline powder for spectroscopy
4-Nitrophenol
p-Nitrophenol is used as an indicator in 0.1% alcohol solutions. 4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidine and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis.
- Presentation: Light Brown Crystalline Powder
- Auto Ignition: 510 °C
- Vapor Pressure: 0.6 mmHg ( 120 °C)
- Refractive Index: 1.61 (Predicted)
- pH: 5.6 colorless -7.6 yellow
- pKa: 7.23 (Predicted)
It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.
Moderately soluble in water; freely soluble in alcohol, chloroform or ether.
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Melting Pt: 105…115 °C Flash Pt: 169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007331 CAS Number: 100-02-7 UN: 1663 ADR: 6.1,III |
Specification Test Results
Identity Test | Passes |
Purity | ≥99% (GC) |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...