L-Methionine ≥99%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid

02102291.1 02102291.3 0210229101
IC102291.1EA 85.65 USD
IC102291.1 IC102291.3 IC10229101
L-Methionine ≥99%, Powder
L-Methionine

Soluble in warm dil alcohol; insoluble in abs. alcohol, ether, petroleum ether, benzene, acetone, HCl.

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
MDL Number: MFCD00063097
CAS Number: 63-68-3

Order Now

Specification Test Results

Synonyms L-2-Amino-4-(methylthio)butanoic acid; 1-2-Amino-4-methyl thiobutanoir acid; (S)-2-Amino-4-(methylmercapto)butyric acid
Form White Crystalline Powder
Storage Store at Room Temperature (15-30 °C).
Loss on Drying ≤0.3%
Optical Rotation +22.4 ° to 24.7 °(c=20, 6N HCl) (Lit.)
pH 5.6 - 6.1 (1% aq soln)
pka 2.28, 9.21 (Lit.)
Residue on Ignition ≤0.4%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR