Menadione (2-methyl-1,4-naphthoquinone, vitamin K3) ≥98%, yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione

0210225925 0210225980
IC10225925EA 83.35 USD
IC10225925 IC10225980
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3) ≥98%, yellow powder
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3)

Menadione belongs to the Vitamin K class of compounds, which are necessary for the biosynthesis of prothrombin and other blood clotting factors. Menadione has demonstrated cytotoxic activity against a variety of cell culture lines and can induce apoptosis of cultured cells, such as osteoclasts and osteoblasts, via elevation of peroxide and superoxide radical levels.


It is a prothrombogenic compound1 and is used as a model quinone in cell culture and in vivo investigations.


Menadione (Vitamin K3) is a synthetic analogue of of 1,4-naphthoquinone with a methyl group in the 2-position. It is used as a phosphatase inhibitor and an inhibitor of mitochondrial DNA polymerase γ (pol γ). It can be used as an oxidative injury (free radical generator) inducing agent.


Soluble in methanol (15 mg/ml-clear solution).


Menadione belongs to the Vitamin K class of compounds, which are necessary for the biosynthesis of prothrombin and other blood clotting factors. Menadione has demonstrated cytotoxic activity against a variety of cell culture lines and can induce apoptosis of cultured cells, such as osteoclasts and osteoblasts, via elevation of peroxide and superoxide radical levels.

Formula: C₁₁H₈O₂
MW: 172.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001681
CAS Number: 58-27-5

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Melting Point 100 - 110°C
Expiration Date Month/Day/Year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR