D-(+)-Mannose ≥98%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal

0210225025 0210225080 0210225090
IC10225025EA 89.2 USD
IC10225025 IC10225080 IC10225090
D-(+)-Mannose ≥98%, white crystalline powder
D-(+)-Mannose

Mannose is a sugar monomer of the aldohexose series of carbohydrates.


D-(+)-Mannose, a C-2 epimer of D-glucose, is used in the formation of glycan structure and glycosylation. D-Mannose has been used in a study to assess the synthesis of a family of amphiphilic glycopolymers. It has also been used in a study to investigate the early detection of bronchiolitis obliterans after lung transplantation.


Soluble in water (50 g/ml - clear colorless solution).


Mannose is a sugar monomer of the aldohexose series of carbohydrates. Mannose is a C-2 epimer of glucose. Mannose is important in human metabolism, especially in the glycosylation of certain proteins. Several congenital disorders of glycosylation are associated with mutations in enzymes involved in mannose metabolism


Precaución: Research Use Only (RUO).

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064122
CAS Number: 3458-28-4

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to standard
Melting Point 129 - 140 °C
Loss on Drying ≤0.5%
Optical Rotation (c=4, H₂O, 24 hour) +14.5±1°
Heavy Metals ≤10 ppm
Residue on Ignition ≤0.1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR