α-Linolenic acid 45 - 65%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid

0210218880 0210218890 0210218891
IC10218880EA 99.28 USD
IC10218880 IC10218890 IC10218891
α-Linolenic acid 45 - 65%, liquid
α-Linolenic acid

Linolenic acid belongs to the class of omega-3 fatty acids.

Formula: C₁₈H₃₀O₂
MW: 278.44 g/mol
MDL Number: MFCD00065720
CAS Number: 463-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR