L(+)-Leucine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

02102158.3 0210215801
IC102158.3EA 40.71 USD
IC102158.3 IC10215801
L(+)-Leucine, white powder
L(+)-Leucine

Leucine is a branched-chain α-amino acid. Leucine is classified as a hydrophobic amino acid due to its aliphatic isobutyl side chain.


Leucine has been used as a molecular marker in the recovery of DNA from palaeontological samples for PCR analysis.


Soluble in water (25 mg/ml); sparingly soluble in alcohol.


Leucine is a branched-chain α-amino acid. Leucine is classified as a hydrophobic amino acid due to its aliphatic isobutyl side chain.


Precaución: Research Use Only (RUO).

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 293 °C
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Specific Rotation +14,5 ° to +16,5 °
Loss on drying ≤0.2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR