Kinetin ≥95%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210211701 0210211705
IC10211701EA 87.63 USD
IC10211701 IC10211705
Kinetin ≥95%, white powder
Kinetin

Kinetin is an adenine-type cytokinin phytohormone.


Kinetin is used as plant growth accelerator. It is used in plant culture media such as Murashige & Skoog media in conjunction with auxins such as indole-3-acetic acid (IAA), Indole-3-butyric acid (IBA), 1-napthaleneacetic acid (NAA) or others to induce the fomation of callus and to regenerate plant tissues from callus. It has been also studied as a treatment for many human disease states caused by pre-mRNA splicing errors, including familial dysautonomia and neurofibromatosis.


Soluble in 0.1N Sodium hydroxide (1% w/v).


Kinetin is an adenine-type cytokinin phytohormone.


Precaución: Research Use Only (RUO).

Formula: C₁₀H₉N₅O
Storage Temperature: Freezer
MDL Number: MFCD00075757
CAS Number: 525-79-1

Order Now

Specification Test Results

Identity Test Passes
Purity ≥95%
FTIR Conforms to Standard
Melting Point 262 - 272 °C
UV/Vis: λmax (ethanol) 267 ± 5 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR