D-(-)-Isoascorbic acid, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210207380 0210207390
IC10207380EA 60.59 USD
IC10207380 IC10207390
D-(-)-Isoascorbic acid, Powder
D(-)-Isoascorbic acid
Formula: C₆H₈O₆
MDL Number: MFCD00005378
CAS Number: 89-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR