4-(Indol-3-yl)butyric acid, off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 3-Indolebutyric acid, Indole-3-butyric acid

0210204201 0210204250
IC10204201EA 48.55 USD
IC10204201 IC10204250
4-(Indol-3-yl)butyric acid, off-white powder
4-(Indol-3-yl)butyric acid

Indole-3-butyric acid is used as plant growth hormone.


A naturally occurring phytohormone auxin (plant growth regulator). Promotes root formation in cuttings, but does not affect ethylene levels.


Indole-3-butyric acid is used as plant growth hormone.


Precaución: Research Use Only (RUO).

Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
MDL Number: MFCD00005664
CAS Number: 133-32-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 118 - 126 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR