Indol-3-acetic acid ~99, off-white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid
Indole-3-acetic acid (IAA) is the most common, naturally-occurring, plant growth regulator - as a principal auxin of higher plants. It is the best known of the auxins, and has been the subject of extensive studies by plant physiologists. Chemically, IAA is a carboxylic acid in which the carboxyl group is attached through a methylene group to the C-3 position of an indole ring.
Indole-3-acetic acid, one of its derivatives bis-2-indolyl disulphide has been used to synthesize tyrosin kinase inhibitors. It can also be used as a starting material to synthesize DBI receptor ligands. Methyl ester of indole-3-acetic acid reacts with dimethylamine and the resulting amide on reduction with Lithium aluminium hydride gives the hallucinogen N,N-Dimethyltryptamine. Used as a plant growth regulator as a principle auxin of higher plants. Can also be used as a starting material to synthesize DBI receptor ligands.
Soluble in methanol, ethanol (5%), Sparingly soluble in water, chloroform. Soluble in acetone, ether and freely soluble in alcohol.
Indole-3-acetic acid (IAA) is the most common, naturally-occurring, plant growth regulator - as a principal auxin of higher plants. It is the best known of the auxins, and has been the subject of extensive studies by plant physiologists. Chemically, IAA is a carboxylic acid in which the carboxyl group is attached through a methylene group to the C-3 position of an indole ring.
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol |
MDL Number:
MFCD00005636 CAS Number: 87-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...