D-(+)-Galactosamine hydrochloride ≥99%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
(2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
D-(+)-Galactosamine hydrochloride is a powerful inhibitor of hepatic RNA synthesis.
D-(+)-Galactosamine (D-chondrosamine) is used with lipopolysaccharides (LPS) to induce models of acute hepatic failure (LPS/D-GalN-induced liver injury, hepatitis) for therapeutic research to find new drugs.
Increases the adherence of polymorphonuclear leukocytes to hepatic endothelial cells and induces superoxide production. Induces hepatic damage, causing macrophage infiltration.
Soluble in water (5% - clear, colourless solution).
Increases the adherence of polymorphonuclear leukocytes to hepatic endothelial cells and induces superoxide production. Induces hepatic damage, causing macrophage infiltration.
Formula:
C₆H₁₄ClNO₅ MW: 215.64 g/mol Melting Pt: 182…185 °C (dec., lit.) |
MDL Number:
MFCD00135830 CAS Number: 1772-03-8 |
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
Optical Rotation (c=1, H2O, after 24 hrs.) | +93 ± 5° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...