4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), tan powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210168505 0210168525 0210168580
IC10168505EA 62.56 USD
IC10168505 IC10168525 IC10168580
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), tan powder
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)

Ferulic acid is a chemopreventive caffeic acid analogue. Ferulic acid is an inhibitor of 5-LO (5-lipoxygenase) and 12-LO (12-lipoxygenase).


Ferulic acid is an antioxidant which neutralizes free radicals (superoxide, nitric oxide and hydroxyl radical) which could cause oxidative damage of cell membranes and DNA.


Soluble in hot water. (Lit.)


Ferulic acid is an antioxidant which neutralizes free radicals (superoxide, nitric oxide and hydroxyl radical) which could cause oxidative damage of cell membranes and DNA.

Formula: C₁₀H₁₀O₄
Melting Pt: 168…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00004400
CAS Number: 1135-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR