Docusate sodium, Solid_waxy_white

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate

0210157550 0210157590
IC10157550EA 64.34 USD
IC10157550 IC10157590
Docusate sodium, Solid_waxy_white
Docusate sodium

Dioctyl sulfosuccinate sodium salt is used to make a microemulsion with CAPSO for the electrophoresis detection of natural and synthetic estrogens. Used to prepare reverse micelles.


Dioctyl sodium sulfosuccinate (DSS) dissolves slowly in water; at 25 °C to the extent of 1.5 gm/100cc; at 70 °C, 5.5 gm/100cc. It dissolves in oils, hydrocarbons, fats and waxs by heating above 75 °C and remains in solution when cooled to room temperature. At room temperature, it is readily soluble in most organic solvents, both polar and non-polar. Soluble in carbon tetrachloride, petroleum ether, naphtha, xylene, dibutyl phthalate, liquid petroleum, acetone, alcohol, vegetable oils.


Dioctyl sulfosuccinate sodium salt is used to make a microemulsion with CAPSO for the electrophoresis detection of natural and synthetic estrogens. Used to prepare reverse micelles.


Precaución: Research Use Only (RUO).

Formula: C₂₀H₃₇NaO₇S
MW: 444.57 g/mol
Melting Pt: 155
MDL Number: MFCD00012455
CAS Number: 577-11-7
Merck Index: 12,03460
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88f50a69ecb539f7bf489da2c2185afda137f66e&vid=efea410db303959215752ea09ff72a4d0b438a78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 5 - 8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR