O-Dianisidine, light brown powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 4,4'-Diamino-3,3'-dimethoxybiphenyl, 3,3'-Dimethoxybenzidine

0210153601 0210153605
IC0210153601EA 44.22 USD
IC0210153601 IC0210153605
O-Dianisidine, light brown powder
O-Dianisidine

o-Dianisidine is a peroxidase substrate suitable for use in ELISA procedures.


  • Presentation: Light Brown Powder
  • Vapor Pressure: 8.8 x 10-9 mm Hg at 25 °C (estimated)


o-Dianisidine is used for the detection of Au, Co, Cu, SCN-, V; determination of Au, NO2-, Ce(IV) (spectrophotometrically). Ortho Dianisidine may be used in a novel automated method, used to measure the total antioxidant response (TAR) against potent free radical reactions. It may also be used along with glucose oxidase and peroxidase in the determination of blood and urinary glucose. o-Dianisidine is used as a redox indicator. It is used as an intermediate for the preparation of azo dyes, pigments and o-dianisidine diisocyanate, which finds application in adhesives, polyurethane elastomers and resins. It is also used in various industries like leather, paper, plastics, rubber, and textiles. It acts as a reagent to detect metals, thiocyanates, and nitrites.


Soluble in ethanol (200 mg/mL - clear, violet-brown solution) or aqueous acids, benzene, ether, chloroform, acetone, most organic solvents and lipids. Almost insoluble in water."


o-Dianisidine is used for the detection of Au, Co, Cu, SCN-, V; determination of Au, NO2-, Ce(IV) (spectrophotometrically).


Precaución: Research Use Only (RUO). Tan powder may darken on storage to a purple color.

Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
Melting Pt: 130…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00008372
CAS Number: 119-90-4
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=199652de8e0fd40254935c87dfcbecaf5113705a&vid=199652de8e0fd40254935c87dfcbecaf5113705a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Melting Point 130 - 140 °C
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR