2'-Deoxycytidine hydrochloride, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210148301 0210148305 0210148310
IC10148301EA 94.18 USD
IC10148301 IC10148305 IC10148310
2'-Deoxycytidine hydrochloride, white powder
2'-Deoxycytidine hydrochloride

Deoxycytidine is a Deoxyribonucleoside, a component of deoxyribonucleic acid. It is similar to the ribonucleoside cytidine, but with one hydroxyl group removed from the 2' position.


Cell Culture


Soluble in water (50mg/mL-clear colorless solution).


Deoxycytidine is a Deoxyribonucleoside, a component of deoxyribonucleic acid. It is similar to the ribonucleoside cytidine, but with one hydroxyl group removed from the 2' position.


Precaución: Research Use Only (RUO).

Formula: C₉H₁₄ClN₃O₄
Storage Temperature: Ambient
MDL Number: MFCD00012840
CAS Number: 3992-42-5

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR