Cytidine free base, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210145905 0210145910 0210145980
IC10145905EA 52.63 USD
IC10145905 IC10145910 IC10145980
Cytidine free base, white powder
Cytidine

Soluble in water.

Formula: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Melting Pt: 208…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00006545
CAS Number: 65-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR