Cystine dihydrochloride, off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: DL-Cystine dihydrochloride

0210145201 0210145205
IC10145201EA 64.93 USD
IC10145201 IC10145205
Cystine dihydrochloride, off-white powder
Cystine dihydrochloride

Precaución: Research Use Only (RUO).

Formula: C₆H₁₄Cl₂N₂O₄S₂
MW: 313.208 g/mol
Storage Temperature: Ambient
CAS Number: 90350-38-2

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR