L(+)-Cysteine hydrochloride monohydrate, crystals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
|
Synonyms:
(R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate
Cysteine is a major biological source of sulfur and is one of the two common sulfur-containing amino acids.
NMDA glutamatergic receptor antagonist.
Freely soluble in water (50 mg/mL).
Cysteine is a major biological source of sulfur and is one of the two common sulfur-containing amino acids.
Formula:
HSCH₂CH(NH₂)COOH·HCl·H₂O MW: 175.64 g/mol |
MDL Number:
MFCD00065606 CAS Number: 7048-04-6 |
Specification Test Results
Synonyms | Cys; Cysteine chlorhydrate; C |
Form | Clear Crystals |
Storage | Store at Room Temperature (15-30 °C). Store Desiccated. |
Moisture Content | 8 - 12% |
Optical Rotation | +2.0° to +7.5°(c = 20, 1N HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...