L(+)-Cysteine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

0210144425 0210144480 0210144490 0210144405
IC10144425EA 68.3 USD
IC10144425 IC10144480 IC10144490 75875-580
L(+)-Cysteine, white powder
L-Cysteine

L-cysteine is used in producing antioxidants. It also promotes healing of severe burns, fat burning and helps build muscle and promotes the activity of white blood cells. It is a precursor in the food, pharmaceutical, and personal care industries.


  • NMDA glutamatergic receptor agonist.


Soluble in water, alcohol, acetic acid, ammonia water; insoluble in ether, acetone, benzene, ethyl acetate, carbon disulfide, carbon tetrachloride


Precaución: Research Use Only (RUO).

Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
Boiling Pt: 254
Melting Pt: 115
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d57c3a3281f85185ee551f2eba43b979bc28713&vid=9d57c3a3281f85185ee551f2eba43b979bc28713&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 213 - 235°C (dec)
Optical Rotation +7 ° to +13 ° (c=2, 1N HCl)
pH 4 - 8 (5% aq soln)
Refractive Index 8.8 ° (C=8, 1mol/L HCl) (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR