CAPS (3-(cyclohexylamino)propanesulfonic acid), white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
CAPS is a zwitterionic biological buffer which has minimal reactivity with enzymes or proteins and minimal salt effects. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. The high pH of this buffer makes it useful for the transfer of proteins with a pI > 8.5.
CAPS has been used to study pH-dependent processes in enzymology and as an eluent buffer component in the separation of basic drugs by HPLC.
ΔpK/ΔT: - 0.0322
Very soluble in water (1 M - clear, colorless solution).
CAPS is a zwitterionic biological buffer which has minimal reactivity with enzymes or proteins and minimal salt effects. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. The high pH of this buffer makes it useful for the transfer of proteins with a pI
Precaución: Research Use Only (RUO).
Formula:
C₆H₁₁NH(CH₂)₃SO₃H |
MDL Number:
MFCD00003837 CAS Number: 1135-40-6 |
|
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
pH (1% aq soln) | 5,0 - 6,3 |
pka | 10,4 (25 °C) (Lit.) |
Refractive Index | 1,52 |
Sterilisation of Solutions | Autoclave |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...