Guanosine-3',5'-cyclicmonophosphoric acid sodium salt ≥95%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium (4aR,6R,7R,7aS)-6-(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)-7-hydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate 2-oxide, Cyclic GMP sodium

0210143325 0210143380 0210143383 0210143390 0210143391
IC10143325EA 61.22 USD
IC10143325 IC10143380 IC10143383 IC10143390 IC10143391
Guanosine-3',5'-cyclicmonophosphoric acid sodium salt ≥95%, Powder
Guanosine-3',5'-cyclicmonophosphoric acid sodium salt

H2O: soluble (50 mg/mL)


An important second messenger, cGMP is a major intracellular mediator of extracellular signals such as nitric oxide and natriuretic peptides. Cyclic GMP interacts with three types of intracellular receptor proteins: cGMP-dependent protein kinases, cGMP-regulated channels, and cGMP-regulated cyclic nucleotide phosphodiesterases. A primary action of elevated cGMP levels in vivo is the stimulation of cGMP-dependent protein kinase (PKG).

Formula: C₁₀H₁₁N₅NaO₇P
MW: 367.19 g/mol
MDL Number: MFCD00038424
CAS Number: 40732-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR