Corn syrup, liquid, MP Biomedicals
Supplier: MP Biomedicals
0210141301
IC10141301EA 123.33 USD
IC10141301
Corn syrup, liquid, MP Biomedicals
Animal Supplies Animal Feed
Corn-syrup is a dual acid-enzyme conversion syrup which contains approximately three times as much maltose as regular corn syrups processed to the same degree of conversion using acid only.
  • Conversion: acid-enzyme
  • Maltose Content: ~30%
Corn syrup is used in commercially prepared foods as a thickener, a sweetener and as a humectant - an ingredient that retains moisture and thus maintains a food's freshness.
Precaución: Research Use Only (RUO).