Cholesterol 99, powder_white

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

0210138201 0210138205 0210138225 0210138280
IC10138201EA 80.91 USD
IC10138201 IC10138205 IC10138225 IC10138280
Cholesterol 99, powder_white
Cholesterol

Cholesterol is a component used for the preparation and study of artificial model membranes. One example is a study of valinomycin induced changes in membrane potentials of red blood cell and phospholipid (phosphatidylcholine from egg yolk plus cholesterol) vesicle suspensions. Positively charged cyanine dyes were used that fluorimetrically respond to the change in potential. Cholesterol is suitable for use in liposome preparations. It is standard for clinical work.


Major component of all biological membranes; ~25% of total brain lipid is cholesterol.


Cholesterol monohydrate is practically insoluble in water (less then 0.5 mg/100 mL of water); slightly soluble in alcohol (1.29% w/w at 20 °C), more soluble in hot alcohol (100 g of saturated 96% alcoholic solution contains 28 g at 80 °C). Also one gram dissolves in 2.8 mL of ether, in 4.5 mL of chloroform, and in 1.5 mL of pyridine. The product is soluble in acetone, dioxane, ethyl acetate, benzene, petroleum ether, oils, fats, and in aqueous solutions of bile salts. Solutions should be protected from light.


Cholesterol is a component used for the preparation and study of artificial model membranes. One example is a study of valinomycin induced changes in membrane potentials of red blood cell and phospholipid (phosphatidylcholine from egg yolk plus cholesterol) vesicle suspensions. Positively charged cyanine dyes were used that fluorimetrically respond to the change in potential. Cholesterol is suitable for use in liposome preparations. It is standard for clinical work.


Precaución: Research Use Only (RUO).

Formula: C₂₇H₄₆O
MW: 386.66 g/mol
Boiling Pt: 360
Storage Temperature: Freezer
MDL Number: MFCD00003646
CAS Number: 57-88-5
Merck Index: 13,02221

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
Loss on Drying ≤1.0%
Optical Rotation -34° to -42°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR