Cesium chloride ≥99.0%, white crystalline powder, Reagent Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Cesium Chloride

0210132125 0210132180 0210132183 0210132190 0210132191
IC10132125EA 84.45 USD
IC10132125 IC10132180 IC10132183 IC10132190 IC10132191
Cesium chloride ≥99.0%, white crystalline powder, Reagent Grade
Cesium chloride

Cesium chloride, an inorganic compound, is an important source of cesium ions in a variety of applications. Its crystal structure forms a major structural type where each cesium ion is coordinated by 8 chlorine ions. Cesium chloride occurs naturally in mineral waters and as an impurity in carnallite (up to 0.002%), sylvite and kainite. Less than 20 tonnes of CsCl is produced annually worldwide, mostly from a cesium-bearing mineral pollucite.


Cesium chloride is typically used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins. It is used for the preparation of electrically conducting glasses, used to make solutions for the separation of RNA from DNA by density gradient centrifugation. Cesium chloride (CsCl) is commonly used in the separation of DNA and RNA. Procedures have been reported for the purification of closed circular DNA by equilibrium centrifugation in both continuous and discontinuous CsCl-ethidium bromide gradients. Subsequently, the CsCl can be removed from the DNA preparation by either dialysis or precipitation.


Cesium chloride is typically used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins. It is used for the preparation of electrically conducting glasses, used to make solutions for the separation of RNA from DNA by density gradient centrifugation. Cesium chloride (CsCl) is commonly used in the separation of DNA and RNA. Procedures have been reported for the purification of closed circular DNA by equilibrium centrifugation in both continuous and discontinuous CsCl-ethidium bromide gradients. Subsequently, the CsCl can be removed from the DNA preparation by either dialysis or precipitation.

Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303 (lit.)
Melting Pt: 646 (lit.)
Density: 3.99 g/mL (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
Merck Index: 13,02021
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eda375136e1b0b37472c813050b7f1f8930242f5&vid=eda375136e1b0b37472c813050b7f1f8930242f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99.0%
pH (1% aq soln) 5.5-6.5
Solubility (50% aq soln) Clear and Colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR