Carboxymethyl cellulose sodium salt, 50 - 200 cps (4% H₂O), powder, low viscosity

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0210127890 0210127891
IC10127890EA 157.25 USD
IC10127890 IC10127891
Carboxymethyl cellulose sodium salt, 50 - 200 cps (4% H₂O), powder, low viscosity
Carboxymethyl cellulose sodium salt

Carboxymethyl ether cellulose sodium salt used as suspending agents. Its viscosity is both concentration and temperature dependent. As the temperaure increases, the viscosity decreases. As the concentration increases, the viscosity increases.


Carboxymethyl ether cellulose is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes, as protective colloid in general. As stabilizer in foods. It is used in pharmaceuticals as a suspending agent, tablet excipient, viscosity-increasing agent. Low viscosity is usually used in thin aqueous solutions.


The solid should be added to the water. Stir gently or shake intermittently; do not stir constantly with a magnetic stirring bar.


Soluble in aqueous solution at 25 °C.


Carboxymethyl ether cellulose sodium salt used as suspending agents. Its viscosity is both concentration and temperature dependent. As the temperaure increases, the viscosity decreases. As the concentration increases, the viscosity increases.

Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number: MFCD00081472
CAS Number: 9004-32-4

Order Now

Specification Test Results

Synonyms CMC; Carboxymethyl ether cellulose sodium salt; Sodium carboxymethylcellulose; Sodium cellulose glycolate; Cellulose glycolic acid sodium salt
Form White Powder
Storage Store at Room Temperature (15-30 °C).
pH 7.6 (4% aq soln) (Lit.)
Viscosity Low Viscocity: 50 - 200 cps (4% aq soln)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR