Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥97%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt
Uridine-5'-Triphosphate Trisodium Salt is a nucleoside diphosphate.
UTP has been used to study its vasodilatory effects on the skeletal muscle cells of diabetic patients[3]. UTP has also been used as a purinergic agonist to study ion transport functions of P2Y2 receptors on rat basolateral membranes.
Soluble in water (50 mg/mL-clear to slightly hazy solution).
Uridine-5'-Triphosphate Trisodium Salt is a nucleoside diphosphate.
Precaución: Research Use Only (RUO).
Formula:
C₉H₁₂N₂Na₃O₁₅P₃ MW: 550.09 g/mol |
MDL Number:
MFCD00044310 CAS Number: 19817-92-6 |
Specification Test Results
Identity Test | Passes |
Purity | ≥97% |
FTIR | Conforms to Standard |
OD 250/260 | 0.74 - 0.79 |
OD 280/260 | 0.32 - 0.40 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...