Disodium UDP-glucose ≥90%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: UDPGUridine-5'-diphosphoglucoseUridine 5′-diphosphoglucose disodium saltUDP-α-D-Glucose disodium saltUDP-GlucoseUridine-5′-diphosphoglucose disodium salt((2R,3S,4R,5R)-5-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl ((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)diphosphate sodium

0210120891 0210120890 0210120894 0210120880
IC0210120894EA 1693.05 USD
IC0210120894 IC10120891 IC10120880 IC10120890
Disodium UDP-glucose ≥90%, white powder
Disodium UDP-glucose

Uridine-5'-diphosphoglucose Disodium Salt is a biosynthetic product produced by the reaction of UTP and glucose-1-phosphate catalyzed by uridyl transferase.


Freely soluble in water; sparingly soluble in methanol; practically insoluble in alcohol, ether, pyridine.


Precaución: Research Use Only (RUO).

Formula: C₁₅H₂₂N₂Na₂O₁₇P₂
MW: 610.26 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077895
CAS Number: 28053-08-9

Order Now

Specification Test Results

Identity Test Passes
Purity ≥90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR