Stearic acid ≥99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Octadecanoic acid, Octadecylic acid

0210111705 0210111710 0210111725 0210111780
IC10111705EA 69.53 USD
IC10111705 IC10111710 IC10111725 IC10111780
Stearic acid ≥99%, white crystalline powder
Stearic acid

Stearic Acid, an anti-inflammatory agent, is the hydrogenation product of linoleic acid.


Stearic acid has been shown to induce a decrease of cell growth and adhesion and increase hydrophobic properties of collagen films. Stearic Acid has been noted to induce an increase in ICAM-1 expression in cells, via an increase in phosphorylation of the transcriptional regulator NFκB


Soluble in alcohol, benzene, chloroform, acetone, carbon tetrachloride, carbondisulfide; very slightly soluble in water.


Stearic Acid, an anti-inflammatory agent, is the hydrogenation product of linoleic acid.

Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
MDL Number: MFCD00002752
CAS Number: 57-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88fb4b039688bfa1460a75817f6fea5b344cfd70&vid=88fb4b039688bfa1460a75817f6fea5b344cfd70&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR