Bicine, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
Bicine is an organic compound used as a buffering agent. It is a zwitterionic buffer and one of the Good's buffers . It is prepared by the reaction ofglycine with ethylene oxide, followed by hydrolysis of the resultant lactone. It is a recommended buffer for low temperature biochemical work. Used for the preparation of stable substrate solution for serum guanase determination.
Bicine is used in a thin layer ion exchange chromatography method for protein resolution.It is used in peptide and protein crystallization.Bicine is used in a kinetic study of a quaternary transition-state analogue complex of creatine kinase and in multiphasic buffer system for SDS-PAGE of proteins.
Soluble in water (10 mg/ml - clear, colourless solution; a saturated solution in water is 1,1 M )
Formula:
C₆H₁₃NO₄ MW: 163.17 g/mol Melting Pt: 190 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00004295 CAS Number: 150-25-4 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Reference |
pH (1% aq soln) | 4.0 - 6.0 |
OD260 nm (10% aq soln) |
|
OD280 nm (10% aq soln) |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...