5-Ethylphenazinium ethyl sulfate ∼95%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: PES, Phenazine ethosulphate

02100956.5 0210095601 0210095605
IC100956.5EA 57.36 USD
IC100956.5 IC10095601 IC10095605
5-Ethylphenazinium ethyl sulfate ∼95%, powder
5-Ethylphenazinium ethyl sulphate

Phenazine ethosulfate is an intermediate which is used to detect nitric oxide reducatase activity in concert with ascorbic acid.


Soluble in water (10 mg/ml - clear, yellow solution) or ethanol (50 mg/ml - clear, dark yellow to orange solution).


Precaución: Research Use Only (RUO).

Formula: C₁₆H₁₈N₂O₄S
MW: 334.4 g/mol
Storage Temperature: Freezer
CAS Number: 10510-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=245e7ef8b3f6136a0840fac5da9356b7dc5ca8fe&vid=245e7ef8b3f6136a0840fac5da9356b7dc5ca8fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ~95%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR